5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)

Supplier: BACHEM AMERICAS INC

4002995.0001 M-1205.0005
M-1205.0001BAEA 495.3 USD
M-1205.0001BA M-1205.0005BA
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)

Sequence: 5-Bromo-4-chloro-1H-indol-3-yl phosphate · p-toluidine


Indoxyl esters and glycosides are often used chromogenic substrates in histochemistry. An indigoid dye is generated by the action of the enzyme. Its absorption maximum is determined by the nature and position of the indoxyl substituents: λmax 605 nm (without substituents), λmax 620 nm (5-bromo-4-chloro), λmax 565 nm (5-bromo-6-chloro).


Synonym(s): BCIP · p-toluidine#X-Phosphate · p-toluidine#Phosphoric acid-mono-(5-bromo-4-chloro-1H-indol-3-yl) ester · p-toluidine

Formula: C₁₅H₁₅BrClN₂O₄P
Storage Temperature: Refrigerator
MDL Number: MFCD00040636
CAS Number: 6578-06-9
EINECS: 229-506-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR