Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser

Supplier: Bachem
4031239.0005 4031239.0001
M-1025.0001BAEA 110.22 USD
M-1025.0001BA M-1025.0005BA
Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser
Sequence: Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser-OH
Synonym(s): Acetyl-Tetradecapeptide Renin Substrate (horse)#Acetyl-Angiotensinogen (1-14) (horse)#Acetyl-Preangiotensinogen (1-14) (horse)#Acetyl Angiotensinogen (1-14), porcine

Renin cleavage of Ac-DRVYIHPFHLLVYS yields N-terminally acetylated angiotensin I and LVYS.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR