Suc-Leu-Leu-Val-Tyr-pNA
Supplier: Bachem
4035739.0025 4035739.0100
L-2130.0025BAEA 579.33 USD
L-2130.0025BA L-2130.0100BA
Suc-Leu-Leu-Val-Tyr-pNA
Proteins and Peptides
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Suc-LLVY-pNA, chromogenic substrate analog to I-1395, Suc-LLVY-AMC.