Glycine-p-nitroanilide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Gly-pNA

4009559.0025 4009559.0005 4009559.0001
L-1280.0025BAEA 618.93 USD
L-1280.0025BA L-1280.0005BA L-1280.0001BA
Glycine-p-nitroanilide
Glycine-p-nitroanilide

Sequence: H-Gly-pNA


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
See also Boc-Gly-ONp (A-1740) and Z-Gly-ONp (M-1250).


Substrate for glycine aminopeptidase.

Formula: C₈H₉N₃O₃
MW: 195,178 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038175
CAS Number: 1205-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR