Boc-Val-Leu-Gly-Arg-pNA

Supplier: BACHEM AMERICAS INC

4009617.0100 4009617.0025
L-1205.0100BAEA 537.12 USD InStock
L-1205.0100BA L-1205.0025BA
Boc-Val-Leu-Gly-Arg-pNA
Proteins and Peptides
Sequence: Boc-Val-Leu-Gly-Arg-pNA
CAS-No.: 68223-95-0
Mol.weight: 663.78
SumFormula: C₃₀H₄₉N₉O₈
Bulk item no.: L-1205
Synonym(s):
EINECS no.: -
MDL no.: MFCD02259646

Boc-VLGR-pNA, chromogenic substrate for horseshoe crab clotting enzyme.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR