BACEJ-1175
4005526.0250 4005526.1000
J-1175.1000BAEA 433.39 USD
J-1175.1000BA J-1175.0250BA
BACEJ-1175
Sequence: (H-Cys-4MβNA)₂
As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.
Substrate for cystyl aminopeptidase (oxytocinase).