Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Lys-Pro-AMC

Supplier: BACHEM AMERICAS INC

I-1745.0050 I-1745.0250
I-1745.0050BAEA 550.89 USD InStock
I-1745.0050BA I-1745.0250BA
H-Lys-Pro-AMC
Proteins and Peptides
Sequence: H-Lys-Pro-AMC
CAS-No.: 133066-53-2
Mol.weight: 400.48
SumFormula: C₂₁H₂₈N₄O₄
Bulk item no.: I-1745
Synonym(s):
EINECS no.: -
MDL no.: MFCD18782514

KP-AMC, substrate for the dipeptidyl aminopeptidase II (DAP II, EC 3.4.14.2), also named dipeptidyl peptidase II (DPP II). KP-AMC is also cleaved by dipeptidyl peptidase IV.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty