Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

H-Ala-Pro-AFC

Supplier: BACHEM AMERICAS INC

I-1680.0050 I-1680.0250
I-1680.0050BAEA 552.12 USD InStock
I-1680.0050BA I-1680.0250BA
H-Ala-Pro-AFC
Proteins and Peptides
Sequence: H-Ala-Pro-AFC
CAS-No.: 125791-92-6
Mol.weight: 397.35
SumFormula: C₁₈H₁₈F₃N₃O₄
Bulk item no.: I-1680
Synonym(s):
EINECS no.: -
MDL no.: MFCD00798773

See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .

Fluorogenic substrate for dipeptidyl peptidase IV (DPP IV) and Xaa-Pro dipeptidase. Substrate for fibroblast activation protein alpha (FAP, also called antiplasmin-cleaving enzyme, APCE).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty