Z-Val-Lys-Met 7-amido-4-methylcoumarin

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Val-Lys-Met-AMC

4017732.0025 4017732.0100
I-1625.0025BAEA 396 USD
I-1625.0025BA I-1625.0100BA
Z-Val-Lys-Met 7-amido-4-methylcoumarin
Z-Val-Lys-Met 7-amido-4-methylcoumarin

Sequence: Z-Val-Lys-Met-AMC


The motif VKM corresponds to APP (669-671). Substrate for Amyloid A4-generating enzymes and cathepsin B. Hook et al. used Z-VKM-MCA to detect β-secretase activity in isolated chromaffin vesicles.

Formula: C₃₄H₄₅N₅O₇S
Storage Temperature: Freezer
MDL Number: MFCD00133589
CAS Number: 141223-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR