Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tos-Gly-Pro-Lys-AMC

Supplier: BACHEM AMERICAS INC

I-1370.0050 I-1370.0250
I-1370.0050BAEA 621.85 USD InStock
I-1370.0050BA I-1370.0250BA
Tos-Gly-Pro-Lys-AMC
Proteins and Peptides
Sequence: Tos-Gly-Pro-Lys-AMC
CAS-No.: 128202-25-5
Mol.weight: 611.72
SumFormula: C₃₀H₃₇N₅O₇S
Bulk item no.: I-1370
Synonym(s):
EINECS no.: -
MDL no.: MFCD00152183

Tos-GPK-AMC, a fluorogenic substrate for trypsin, gingipain K, and cathepsin L.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty