H-Ser-AMC

Supplier: BACHEM AMERICAS INC

4002585.1000 4002585.0250
I-1305.1000BAEA 2973.42 USD
I-1305.1000BA I-1305.0250BA
H-Ser-AMC
H-Ser-AMC

Sequence: H-Ser-AMC


Bachem also offers derivatives of phosphoserine and sulfoserine.
See also H-Ser(Ac)-OH, E-1200.

Formula: C₁₃H₁₄N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00079565
CAS Number: 98516-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR