Glu-NH-med

Supplier: BACHEM AMERICAS INC

Synonyms: H-Glu-AMC

4002702.0250
I-1180.0250BAEA 855 USD
I-1180.0250BA
Glu-NH-med
Glu-NH-med

Sequence: H-Glu-AMC


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
See also the glutamic acid γ-diazomethyl ketones L-DON (F-1510) and D-DON (F-2185).


Substrate for aminopeptidase A.

Formula: C₁₅H₁₆N₂O₅
MW: 304.302 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00051217
CAS Number: 98516-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR