ZINC71788382

Supplier: BACHEM AMERICAS INC

Synonyms: H-Ala-Phe-Lys-AMC

4005499.0050
I-1045.0050BAEA 794.04 USD
I-1045.0050BA
ZINC71788382
ZINC71788382

Sequence: H-Ala-Phe-Lys-AMC


AFK-AMC, fluorogenic substrate for a simple and direct assay of plasmin, even in the presence of urokinase and thrombin.

Formula: C₂₈H₃₅N₅O₅
MW: 521.618 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038709
CAS Number: 120928-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR