H-D-Ala-AMC·TFA

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Ala-AMC·TFA(R)-2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)propanamide 2,2,2-trifluoroacetate

4006785.0250
I-1025.0250BAEA 701.03 USD
I-1025.0250BA
H-D-Ala-AMC·TFA
H-D-Ala-AMC·TFA

Sequence: H-D-Ala-AMC · TFA


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
See also Z-Ala-ONp, C-1040.

Formula: C₁₅H₁₅F₃N₂O₅
MW: 360.289 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151971
CAS Number: 201847-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR