BACEI-1000
4000973.0050 4000973.0250
I-1000.0050BAEA 644.92 USD
I-1000.0050BA I-1000.0250BA
BACEI-1000
Sequence: Ac-Ala-Ala-Pro-Ala-AMC
See also I-1270, L-1000, L-1090, L-1180, L-1335, L-1385, L-1400, M-1170, and M-1250.
A specific fluorogenic substrate for human leukocyte and especially hog pancreatic elastases. Ac-AAPA-AMC has been used by Konno et al. for crystalizing E.coli cyclophilin B.