N-α-Cbz-L-ornithine

Supplier: BACHEM AMERICAS INC

Synonyms: H-DL-Orn(Z)-OH

4039043.0005
F-4075.0005BAEA 228.59 USD
F-4075.0005BA
N-α-Cbz-L-ornithine
N-alpha-Cbz-L-ornithine

Sequence: H-DL-Orn(Z)-OH

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210831
CAS Number: 70671-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR