H-D-Dap(Boc)-OMe

Synonyms: H-D-Dap(Boc)-OMe·HCl

4038071.0001 4038071.0005
F-4065.0005BAEA 1269.05 USD
F-4065.0005BA F-4065.0001BA
H-D-Dap(Boc)-OMe
H-D-Dap(Boc)-OMe

Sequence: H-D-Dap(Boc)-OMe · HCl


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.

Formula: C₉H₁₈N₂O₄
MW: 218.253 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05663450
CAS Number: 363191-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR