BACEF-4060

4038482.0005 4038482.0025
F-4060.0025BAEA 1708.76 USD
F-4060.0025BA F-4060.0005BA
BACEF-4060
Sequence: D-1,2,3,4-Tetrahydronorharman-3-carboxylic acid

Tpi, a constrained Trp analog, is also called 1,2,3,4-Tetrahydro-β-carboline-3-carboxylic acid.

Synonym(s): H-D-Tpi-OH#D-1,2,3,4-Tetrahydro-β-carboline-3-carboxylic acid#D-Tryptoline-3-carboxylic acid#D-1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole-3-carboxylic acid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR