Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2-Amino-2-phenylacetamide

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Phg-NH₂·HClD(-)-PhenylglycinamideH-D-Phg-NH2

F-3895.0001 F-3895.0005
F-3895.0001BAEA 85.73 USD InStock
F-3895.0001BA F-3895.0005BA
(R)-2-Amino-2-phenylacetamide
(R)-2-Amino-2-phenylacetamide
Sequence: H-D-Phg-NH₂ · HCl
CAS-No.: 6485-67-2
Mol.weight: 186.64
SumFormula: C₈H₁₀N₂O · HCl
Bulk item no.: F-3895
Synonym(s):
EINECS no.: 420-370-0
MDL no.: MFCD01863288

CAS Number (hydrochloride): 63291-39-4.


Formula: C₈H₁₀N₂O
MW: 150,181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06799064
CAS Number: 6485-67-2
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty