(R)-2-amino-2,3-dimethylbutanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-α-Me-D-Val-OH(R)-2-Amino-2,3-dimethylbutanoic acid

4026441.0001
F-3540.0001BAEA 533.27 USD
F-3540.0001BA
(R)-2-amino-2,3-dimethylbutanoic acid
(R)-2-amino-2,3-dimethylbutanoic acid

Sequence: H-α-Me-D-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
MeVal, αMeVal.

Formula: C₆H₁₃NO₂
MW: 131.175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318765
CAS Number: 53940-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR