(H-D-Cys-OMe)₂·2 HCl

4026225.1000
F-3345.1000BAEA 599.05 USD
F-3345.1000BA
(H-D-Cys-OMe)₂·2 HCl
(H-D-Cys-OMe)₂·2 HCl

Sequence: (H-D-Cys-OMe)₂ · 2 HCl


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.

Formula: C₈H₁₈Cl₂N₂O₄S₂
Storage Temperature: Ambient
MDL Number: MFCD09951837
CAS Number: 144000-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR