C14H19NO2·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid-t-butyl ester·HCl

4025890.1000 4025890.0250
F-3310.1000BAEA 489.37 USD InStock
F-3310.1000BA F-3310.0250BA
C14H19NO2·HCl
C14H19NO2·HCl

Sequence: D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid-t-butyl ester · HCl


Tic represents a constrained Phe analog.


Synonym(s): H-D-Tic-OtBu · HCl

Formula: C₁₄H₂₀ClNO₂
MW: 269.769 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672372
CAS Number: 103733-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR