α-Methyl-D-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-α-Me-D-Phe-OH(R)-(+)-2-Amino-2-methyl-3-phenylpropionic acid(R)-ɑ-Methylphenylalanine(R)-2-Amino-2-methyl-3-phenylpropanoic acidα-Me-D-Phe-OH

4025136.0001
F-3115.0001BAEA 380.9 USD
F-3115.0001BA
α-Methyl-D-phenylalanine
α-Methyl-D-phenylalanine

Sequence: H-α-Me-D-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₁₀H₁₃NO₂
MW: 179.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153491
CAS Number: 17350-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR