Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Methionine

Supplier: BACHEM AMERICAS INC

Synonyms: Methionine racematic(±)-2-Amino-4-(methylthio)butanoic acid(±)-2-Amino-4-(methylthio)butyric acid(±)-MethionineH-DL-Met-OHMethioninum racemicum

F-2910.0100 F-2910.0025
F-2910.0100BAEA 162.53 USD InStock
F-2910.0100BA F-2910.0025BA
DL-Methionine
DL-Methionine
Sequence: H-DL-Met-OH

Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: ~ 136 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063096
CAS Number: 59-51-8
EINECS: 200-432-1
Merck Index: 13,06004
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty