BACEF-2860

4014117.0001 4014117.0005
F-2860.0001BAEA 187.98 USD
F-2860.0001BA F-2860.0005BA
BACEF-2860
Sequence: H-D-His(1-Trt)-OH

The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Synonym(s): H-D-His(τ-Trt)-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR