BACEF-2770
4017506.0001 4017506.0005
F-2770.0005BAEA 397.47 USD
F-2770.0005BA F-2770.0001BA
BACEF-2770
Sequence: H-D-Cys(Trt)-OH
As its L-isomer STLC, D-Cys(Trt) (STDC, NSC 124676) is a reversible, tight binding inhibitor of the human kinesin Eg5 that specifically blocks mitotic progression.