H-D-His(3-Me)-OH
Supplier: BACHEM AMERICAS INC
4017900.1000
F-2600.1000BAEA
716.31
USD
F-2600.1000BA
H-D-His(3-Me)-OH
H-D-His(3-Me)-OH
Sequence: H-D-His(3-Me)-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Synonym(s): H-D-His(π-Me)-OH
Formula:
C₇H₁₁N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00067633 CAS Number: 163750-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...