H-ALLO-Thr(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: H-allo-Thr(tBu)-OHAllo-Thr(tBu)-OH

4016122.0001
F-2540.0001BAEA 489.24 USD
F-2540.0001BA
H-ALLO-Thr(tBu)-OH
H-ALLO-Thr(tBu)-OH

Sequence: H-allo-Thr(tBu)-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₈H₁₇NO₃
MW: 175.228 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237373
CAS Number: 201353-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR