2-Amino-3-fluoropropanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-β-Fluoro-DL-Ala-OH

4015721.0250
F-2530.0250BAEA 502.91 USD
F-2530.0250BA
2-Amino-3-fluoropropanoic acid
2-Amino-3-fluoropropanoic acid

Sequence: H-β-Fluoro-DL-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines

Formula: C₃H₆FNO₂
MW: 107.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077524
CAS Number: 16652-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR