4-Chloro-D-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Phe(4-Cl)-OHH-p-Chloro-D-Phe-OH(R)-2-Amino-3-(4-chlorophenyl)propionic acid

4011117.0001
F-2520.0001BAEA 167.67 USD
F-2520.0001BA
4-Chloro-D-phenylalanine
4-Chloro-D-phenylalanine

Sequence: H-p-Chloro-D-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Synonym(s): (R)-2-Amino-3-(4-chloro-phenyl)-propionic acid#H-4-Chloro-D-Phe-OH#H-D-4Cpa-OH

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Storage Temperature: Ambient
CAS Number: 14091-08-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR