BACEF-2365
4014104.0100 4014104.0025
F-2365.0025BAEA 123.81 USD
F-2365.0025BA F-2365.0100BA
BACEF-2365
Sequence: For-DL-Met-OH
Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.