N-Acetyl-D-methionine
Supplier: BACHEM AMERICAS INC
Synonyms:
Ac-D-Met-OH
4007232.0005
F-2300.0005BAEA
156.24
USD
F-2300.0005BA
N-Acetyl-D-methionine
N-Acetyl-D-methionine
Sequence: Ac-D-Met-OH
Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.
Formula:
C₇H₁₃NO₃S MW: 191.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069962 EINECS: 216-144-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...