3-Fluoro-DL-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: 2-Amino-3-(3-fluorophenyl)propionic acidH-DL-Phe(3-F)-OH

4011126.0005
F-2135.0005BAEA 213.35 USD
F-2135.0005BA
3-Fluoro-DL-phenylalanine
3-Fluoro-DL-phenylalanine

Sequence: H-m-Fluoro-DL-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₉H₁₀FNO₂
MW: 183.2 g/mol
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR