BACEF-1780
4001904.0001 4001904.0005
F-1780.0005BAEA 1425.21 USD
F-1780.0005BA F-1780.0001BA
BACEF-1780
Sequence: N-Me-p-nitro-Phe-OH
Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
p-Nitrophenylalanine (4-Nph, Phe(4-NO₂)) is incorporated into peptides as a chromophore. In FRET substrates, it acts as quencher. Bachem also offers resin-bound Fmoc-Nph (Fmoc-p-nitro-Phe-Wang resin, D-1135).