BACEF-1620

4005684.0005 4005684.0025
F-1620.0005BAEA 173.35 USD
F-1620.0005BA F-1620.0025BA
BACEF-1620
Sequence: H-DL-Homophe-OH

Hph derivatives. Bachem also offers Fmoc-Hph-Wang resin, D-2005.

Synonym(s): (RS)-2-Amino-4-phenyl-butyric acid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR