γ-Carboxyglutamate

Supplier: BACHEM AMERICAS INC

Synonyms: H-γ-Carboxy-DL-Glu-OH

4001775.0010
F-1390.0010BAEA 109.49 USD
F-1390.0010BA
γ-Carboxyglutamate
gamma-Carboxyglutamate

Sequence: H-γ-Carboxy-DL-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): H-DL-Gla-OH

Formula: C₆H₉NO₆
MW: 191.139 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065488
CAS Number: 56271-99-9
EINECS: 260-087-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR