BACEF-1080

4010416.0005 4010416.0001
F-1080.0005BAEA 458.15 USD
F-1080.0005BA F-1080.0001BA
BACEF-1080
Sequence: Ac-DL-Pra-OEt
Synonym(s): Ac-propargyl-DL-Gly-OEt#(RS)-2-Acetylamino-4-pentynoic acid-ethyl ester

Bachem also offers resin-bound propargylglycine, Fmoc-Pra-Wang resin (D-2820).

Precursor for tritiated norleucine.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR