BACEE-3690

4047561.0005 4047561.0025
E-3690.0025BAEA 1239.48 USD
E-3690.0025BA E-3690.0005BA
BACEE-3690
Sequence: H-Aib-OEt · HCl
Synonym(s): H-α-Me-Ala-OEt · HCl
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR