H-Glu(OtBu)-allyl ester · HCl

Supplier: BACHEM AMERICAS INC

4029296.0001
E-3585.0001BAEA 609.62 USD
E-3585.0001BA
H-Glu(OtBu)-allyl ester · HCl
H-Glu(OtBu)-allyl ester · HCl

Sequence: H-Glu(OtBu)-allyl ester · HCl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₂H₂₁NO₄·HCl
Storage Temperature: Freezer
MDL Number: MFCD03093487
CAS Number: 1009669-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR