Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biocytinamide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Lys(biotinyl)-NH₂

E-3430.0250 E-3430.1000
E-3430.0250BAEA 477.44 USD InStock
E-3430.0250BA E-3430.1000BA
Biocytinamide
Biocytinamide
Sequence: H-Lys(biotinyl)-NH₂

For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Bachem also offers resins preloaded with Nε-labeled Fmoc-lysine: Fmoc-Lys(biotinyl)-Wang resin (D-2705) and Fmoc-Lys(Dnp)-SASRIN resin (D-2450).

Synonym(s): Biocytin amide


Formula: C₁₆H₂₉N₅O₃S
MW: 371.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01319117
CAS Number: 61125-53-9
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty