H-Lys(isopropyl)-OH

Supplier: BACHEM AMERICAS INC

4025370.0001
E-3395.0001BAEA 701.03 USD
E-3395.0001BA
H-Lys(isopropyl)-OH
H-Lys(isopropyl)-OH

Sequence: H-Lys(isopropyl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₉H₂₀N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD02259512
CAS Number: 5977-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR