H-DAB(boc)-OMe·HCl

Synonyms: (S)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochlorideH-Dab(Boc)-OMe·HCl

4026986.0005 4026986.0001
E-3360.0005BAEA 868.08 USD
E-3360.0005BA E-3360.0001BA
H-DAB(boc)-OMe·HCl
H-DAB(boc)-OMe·HCl

Sequence: H-Dab(Boc)-OMe · HCl


CAS number (net): 125279-44-9.

Formula: C₁₀H₂₁ClN₂O₄
MW: 268.738 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798680
CAS Number: 3350-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR