H-Asp(OtBu)-allyl ester·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Asp(OtBu)-allyl ester·HCl

4026778.0001 4026778.0005
E-3355.0001BAEA 143.22 USD InStock
E-3355.0001BA E-3355.0005BA
H-Asp(OtBu)-allyl ester·HCl
H-Asp(OtBu)-allyl ester·HCl

Sequence: H-Asp(OtBu)-allyl ester · HCl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₁H₂₀ClNO₄
MW: 265,734 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798679
CAS Number: 218938-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR