H-Hyp(BZL)-OMe·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Hyp(Bzl)-OMe·HCl

4026615.0025 4026615.0001 4026615.0005
E-3315.0025BAEA 799.71 USD InStock
E-3315.0025BA E-3315.0001BA E-3315.0005BA
H-Hyp(BZL)-OMe·HCl
H-Hyp(BZL)-OMe·HCl

Sequence: H-Hyp(Bzl)-OMe · HCl


Hyp denotes L-trans-hydroxyproline.

Formula: C₁₃H₁₈ClNO₃
MW: 271,741 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798674
CAS Number: 66831-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR