Leucine allyl ester

Supplier: BACHEM AMERICAS INC

Synonyms: H-Leu-allyl ester·p-tosylate

4025886.0005
E-3220.0005BAEA 489.24 USD
E-3220.0005BA
Leucine allyl ester
Leucine allyl ester

Sequence: H-Leu-allyl ester · p-tosylate


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672721
CAS Number: 45012-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR