H-Cys(SO₃H)-OH · sodium salt

Supplier: BACHEM AMERICAS INC

Synonyms: Sodium (R)-S-(2-amino-2-carboxyethyl) sulphurothioateSodium (R)-S-(2-amino-2-carboxyethyl) sulfurothioate

4016588.0005 4016588.0025
E-3130.0005BAEA 190.38 USD
E-3130.0005BA E-3130.0025BA
H-Cys(SO₃H)-OH · sodium salt
H-Cys(SO₃H)-OH · sodium salt

Sequence: H-Cys(SO₃H)-OH · sodium salt


A very potent NMDA receptor agonist. Elevated urinary S-sulfocysteine is a metabolic marker of sulfite oxidase deficiency.

Formula: C₃H₆NNaO₅S₂
MW: 223.21 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236947
CAS Number: 7381-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR