AC-Tyr(tBu)-OH

Synonyms: Ac-Tyr(tBu)-OH

4018545.0005
E-3055.0005BAEA 803.6 USD
E-3055.0005BA
AC-Tyr(tBu)-OH
AC-Tyr(tBu)-OH

Sequence: Ac-Tyr(tBu)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236772
CAS Number: 201292-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR