(S)-2-Acetamido-3-(1-trityl-1H-imidazol-4-yl)propanoic acid

Synonyms: Ac-His(1-Trt)-OHAc-His(Trt)-OH

4018523.0001 4018523.0005
E-3045.0005BAEA 803.6 USD
E-3045.0005BA E-3045.0001BA
(S)-2-Acetamido-3-(1-trityl-1H-imidazol-4-yl)propanoic acid
(S)-2-Acetamido-3-(1-trityl-1H-imidazol-4-yl)propanoic acid

Sequence: Ac-His(1-Trt)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Ac-His(τ-Trt)-OH

Formula: C₂₇H₂₅N₃O₃
MW: 439.515 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236758
CAS Number: 183498-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR