BACEE-2945
4011705.0005 4011705.0001
E-2945.0005BAEA 569.57 USD
E-2945.0005BA E-2945.0001BA
BACEE-2945
Sequence: H-Cys(Dpm)-OH
The S-diphenylmethyl (Dpm) group is cleaved with TFA in the presence of phenol.