(H-Cys-OtBu)₂·2 HCl

Supplier: BACHEM AMERICAS INC

Synonyms: (2R,2'R)-Di-tert-butyl 3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride(2R,2'R)-Di-tert-butyl 3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride

4014989.0025 4014989.0005
E-2855.0025BAEA 954.01 USD
E-2855.0025BA E-2855.0005BA
(H-Cys-OtBu)₂·2 HCl
(H-Cys-OtBu)₂·2 HCl

Sequence: (H-Cys-OtBu)₂ · 2 HCl


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.

Formula: C₁₄H₃₀Cl₂N₂O₄S₂
MW: 425.424 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237452
CAS Number: 38261-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR